Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42744649-42745198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

SNP Detail

SNP ID Chromosome Location
rs560041206 Chr.15: 42744690-42744690 on Build GRCh38
rs928031530 Chr.15: 42745100-42745100 on Build GRCh38
rs1036571575 Chr.15: 42744779-42744783 on Build GRCh38
rs532343878 Chr.15: 42744775-42744775 on Build GRCh38
rs890221979 Chr.15: 42744871-42744871 on Build GRCh38
rs536322846 Chr.15: 42745112-42745112 on Build GRCh38
rs34113530 Chr.15: 42744758-42744759 on Build GRCh38
rs902981748 Chr.15: 42744897-42744897 on Build GRCh38
rs1038682016 Chr.15: 42744906-42744906 on Build GRCh38
rs934539405 Chr.15: 42744842-42744842 on Build GRCh38
rs991532320 Chr.15: 42745111-42745111 on Build GRCh38
rs1013224044 Chr.15: 42745046-42745046 on Build GRCh38
rs1035802639 Chr.15: 42744691-42744691 on Build GRCh38
rs561818658 Chr.15: 42744800-42744800 on Build GRCh38
rs957239867 Chr.15: 42744943-42744943 on Build GRCh38
rs1022902264 Chr.15: 42745051-42745054 on Build GRCh38
rs1040094546 Chr.15: 42744884-42744884 on Build GRCh38
rs1052013607 Chr.15: 42744851-42744851 on Build GRCh38
rs368522546 Chr.15: 42744720-42744720 on Build GRCh38
rs567667407 Chr.15: 42744948-42744948 on Build GRCh38
rs769283772 Chr.15: 42745051-42745051 on Build GRCh38
rs994907506 Chr.15: 42744949-42744949 on Build GRCh38
rs1008546636 Chr.15: 42744859-42744859 on Build GRCh38
rs78237254 Chr.15: 42745117-42745117 on Build GRCh38
rs943092788 Chr.15: 42744898-42744898 on Build GRCh38
rs915881004 Chr.15: 42744807-42744807 on Build GRCh38
rs1027270353 Chr.15: 42745032-42745032 on Build GRCh38
rs137990288 Chr.15: 42744889-42744889 on Build GRCh38
rs181464499 Chr.15: 42745126-42745126 on Build GRCh38
rs541966678 Chr.15: 42744681-42744681 on Build GRCh38
rs924516573 Chr.15: 42744831-42744831 on Build GRCh38
rs962081721 Chr.15: 42745105-42745105 on Build GRCh38
rs886051160 Chr.15: 42745104-42745105 on Build GRCh38
rs960088868 Chr.15: 42744765-42744765 on Build GRCh38
rs569220743 Chr.15: 42744828-42744828 on Build GRCh38
rs887341765 Chr.15: 42745049-42745049 on Build GRCh38
rs145576687 Chr.15: 42744829-42744829 on Build GRCh38
rs960121592 Chr.15: 42745086-42745086 on Build GRCh38
rs748831261 Chr.15: 42745057-42745057 on Build GRCh38
rs75447904 Chr.15: 42745116-42745116 on Build GRCh38
rs1033226876 Chr.15: 42744921-42744921 on Build GRCh38

View Full Product Details