Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42751893-42752401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6810859 c.2979C>T p.G993G Chr.15: 42752267-42752267 on Build GRCh38
79082 c.3176G>A p.W1059* Chr.15: 42752070-42752070 on Build GRCh38
5623135 c.3176G>T p.W1059L Chr.15: 42752070-42752070 on Build GRCh38
1629562 c.3212C>T p.S1071F Chr.15: 42752034-42752034 on Build GRCh38
4570689 c.3068T>G p.F1023C Chr.15: 42752178-42752178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765811535 Chr.15: 42752367-42752367 on Build GRCh38
rs370550891 Chr.15: 42752282-42752282 on Build GRCh38
rs373433433 Chr.15: 42752273-42752273 on Build GRCh38
rs756527828 Chr.15: 42752215-42752215 on Build GRCh38
rs779236069 Chr.15: 42752236-42752236 on Build GRCh38
rs776495580 Chr.15: 42752020-42752020 on Build GRCh38
rs757518859 Chr.15: 42752363-42752363 on Build GRCh38
rs766921712 Chr.15: 42752327-42752327 on Build GRCh38
rs759799761 Chr.15: 42751999-42751999 on Build GRCh38
rs376827686 Chr.15: 42752109-42752109 on Build GRCh38
rs770922979 Chr.15: 42752302-42752302 on Build GRCh38
rs978575506 Chr.15: 42752039-42752039 on Build GRCh38
rs969467277 Chr.15: 42752287-42752287 on Build GRCh38
rs903031654 Chr.15: 42752209-42752209 on Build GRCh38
rs770974842 Chr.15: 42751994-42751994 on Build GRCh38
rs955867545 Chr.15: 42752086-42752086 on Build GRCh38
rs987659023 Chr.15: 42752117-42752117 on Build GRCh38
rs758922700 Chr.15: 42752198-42752198 on Build GRCh38
rs778104862 Chr.15: 42752221-42752221 on Build GRCh38
rs201662235 Chr.15: 42752225-42752225 on Build GRCh38
rs911630565 Chr.15: 42752165-42752165 on Build GRCh38
rs779054306 Chr.15: 42752087-42752087 on Build GRCh38
rs771064990 Chr.15: 42752081-42752081 on Build GRCh38
rs972275430 Chr.15: 42752355-42752355 on Build GRCh38
rs766166040 Chr.15: 42752169-42752169 on Build GRCh38
rs55919340 Chr.15: 42752207-42752207 on Build GRCh38
rs989548905 Chr.15: 42751920-42751920 on Build GRCh38
rs745988269 Chr.15: 42752253-42752253 on Build GRCh38
rs761646806 Chr.15: 42752021-42752021 on Build GRCh38
rs34348991 Chr.15: 42751995-42751995 on Build GRCh38
rs55735086 Chr.15: 42752186-42752186 on Build GRCh38
rs55833708 Chr.15: 42752061-42752061 on Build GRCh38
rs139075127 Chr.15: 42751934-42751934 on Build GRCh38
rs752316838 Chr.15: 42752211-42752211 on Build GRCh38
rs372057748 Chr.15: 42751927-42751927 on Build GRCh38
rs750352266 Chr.15: 42752036-42752036 on Build GRCh38
rs879080176 Chr.15: 42752329-42752329 on Build GRCh38
rs778205234 Chr.15: 42751974-42751974 on Build GRCh38
rs570976750 Chr.15: 42752019-42752019 on Build GRCh38
rs772205818 Chr.15: 42751988-42751988 on Build GRCh38
rs757774197 Chr.15: 42752232-42752232 on Build GRCh38
rs371516556 Chr.15: 42752032-42752032 on Build GRCh38
rs928132500 Chr.15: 42751956-42751957 on Build GRCh38
rs760324636 Chr.15: 42752346-42752346 on Build GRCh38
rs1022933283 Chr.15: 42752364-42752364 on Build GRCh38
rs780651650 Chr.15: 42752048-42752048 on Build GRCh38
rs372562825 Chr.15: 42752063-42752063 on Build GRCh38
rs754664311 Chr.15: 42752047-42752047 on Build GRCh38
rs759127398 Chr.15: 42752320-42752320 on Build GRCh38
rs765923162 Chr.15: 42752349-42752349 on Build GRCh38
rs377620628 Chr.15: 42752266-42752266 on Build GRCh38
rs765294536 Chr.15: 42752030-42752030 on Build GRCh38
rs866877821 Chr.15: 42752350-42752350 on Build GRCh38
rs370728842 Chr.15: 42752267-42752267 on Build GRCh38
rs770298532 Chr.15: 42751960-42751960 on Build GRCh38
rs772357636 Chr.15: 42752095-42752095 on Build GRCh38
rs377763812 Chr.15: 42752049-42752049 on Build GRCh38
rs754220583 Chr.15: 42752357-42752357 on Build GRCh38
rs180791005 Chr.15: 42752305-42752305 on Build GRCh38
rs1004340277 Chr.15: 42751937-42751937 on Build GRCh38
rs749779791 Chr.15: 42752077-42752077 on Build GRCh38
rs769692778 Chr.15: 42752134-42752134 on Build GRCh38
rs762984697 Chr.15: 42752155-42752155 on Build GRCh38
rs773343074 Chr.15: 42752153-42752153 on Build GRCh38
rs745874156 Chr.15: 42752091-42752091 on Build GRCh38
rs767231995 Chr.15: 42752203-42752203 on Build GRCh38
rs758376275 Chr.15: 42752258-42752258 on Build GRCh38
rs750869234 Chr.15: 42752370-42752370 on Build GRCh38
rs761736684 Chr.15: 42752124-42752124 on Build GRCh38
rs748395736 Chr.15: 42751959-42751959 on Build GRCh38
rs538954156 Chr.15: 42752045-42752045 on Build GRCh38
rs375008079 Chr.15: 42752037-42752037 on Build GRCh38
rs775188866 Chr.15: 42752337-42752337 on Build GRCh38

View Full Product Details