The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178093.1 | NP_001171564.1 | 3 - 4 | 400 | 63 |
NM_001178094.1 | NP_001171565.1 | 3 - 4 | 697 | 63 | |
NM_005504.6 | NP_005495.2 | 3 - 4 | 804 | 63 | |
GenBank mRNA | AK056255.1 | - | 3 - 4 | 804 | 63 |
AK294879.1 | - | 3 - 4 | 383 | 63 | |
BX647122.1 | - | 3 - 4 | 406 | 63 | |
DC385021.1 | - | 3 - 4 | 401 | 63 | |
U21551.1 | - | 2 - 3 | 271 | 63 |