Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOSTDC1
Entrez Gene ID
25928
Gene Name
sclerostin domain containing 1
Gene Aliases
CDA019, DAND7, ECTODIN, USAG1
Chromosome Location
Chr.7: 16461481 - 16496513 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017011943.1 XP_016867432.1 1 - 2 133 113
GenBank mRNA AK093408.1 - 2 - 3 523 113

View Full Product Details