The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293083.1 | NP_001280012.1 | 49 - 50 | 5593 | 112 |
XM_011512110.2 | XP_011510412.1 | 49 - 50 | 5637 | 112 | |
XM_011512111.2 | XP_011510413.1 | 49 - 50 | 5634 | 112 | |
XM_011512113.2 | XP_011510415.1 | 49 - 50 | 5631 | 112 | |
XM_011512114.2 | XP_011510416.1 | 49 - 50 | 5607 | 112 | |
XM_011512115.2 | XP_011510417.1 | 48 - 49 | 5568 | 112 | |
XM_011512116.2 | XP_011510418.1 | 49 - 50 | 5550 | 112 | |
XM_011512117.2 | XP_011510419.1 | 48 - 49 | 5532 | 112 | |
XM_011512118.2 | XP_011510420.1 | 48 - 49 | 5517 | 112 | |
XM_011512119.2 | XP_011510421.1 | 48 - 49 | 5475 | 112 | |
XM_011512120.2 | XP_011510422.1 | 48 - 49 | 5528 | 112 | |
XM_011512122.2 | XP_011510424.1 | 49 - 50 | 5638 | 112 | |
XM_011512124.2 | XP_011510426.1 | 33 - 34 | 5221 | 112 | |
XM_017005222.1 | XP_016860711.1 | 49 - 50 | 5634 | 112 | |
XM_017005223.1 | XP_016860712.1 | 32 - 33 | 4285 | 112 | |
XM_017005224.1 | XP_016860713.1 | 23 - 24 | 3077 | 112 | |
GenBank mRNA | AL133022.1 | - | 31 - 32 | 4996 | 112 |
AY461813.1 | - | 48 - 49 | 5619 | 112 | |
BC117324.1 | - | 15 - 16 | 1801 | 112 | |
BC126368.1 | - | 15 - 16 | 1804 | 112 | |
BC143825.1 | - | 15 - 16 | 1801 | 112 |