The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206920.2 | NP_996803.2 | 18 - 19 | 2374 | 58 |
GenBank mRNA | AL137659.1 | - | 17 - 18 | 2917 | 58 |
AL834531.1 | - | 3 - 4 | 389 | 58 | |
AY358419.1 | - | 18 - 19 | 2374 | 58 |