The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206920.2 | NP_996803.2 | 24 - 25 | 3135 | 63 |
GenBank mRNA | AL137659.1 | - | 22 - 23 | 3587 | 63 |
AL834531.1 | - | 6 - 7 | 1512 | 63 | |
AY358419.1 | - | 24 - 25 | 3135 | 63 | |
BF339007.1 | - | 5 - 6 | 520 | 63 |