Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
Chromosome Location
Chr.11: 35431827 - 35530300 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001991.2 NP_001001991.1 7 - 8 1477 64
NM_001282675.1 NP_001269604.1 9 - 10 1467 64
NM_001282676.1 NP_001269605.1 5 - 6 1144 64
NM_015430.3 NP_056245.2 8 - 9 1528 64
GenBank mRNA AF370388.1 - 6 - 7 966 64
AK027841.1 - 8 - 9 1128 64
AK300185.1 - 5 - 6 729 64
AK316188.1 - 5 - 6 716 64
AL832391.1 - 7 - 8 1043 64
AY358346.1 - 7 - 8 1056 64
BC089434.1 - 8 - 9 1120 64
BX640676.1 - 7 - 8 1477 64

View Full Product Details