The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321300.1 | NP_001308229.1 | 3 - 4 | 1242 | 109 |
NM_001321303.1 | NP_001308232.1 | 2 - 3 | 1838 | 109 | |
NM_001321304.1 | NP_001308233.1 | 2 - 3 | 1838 | 109 | |
NM_001321305.1 | NP_001308234.1 | 2 - 3 | 1838 | 109 | |
NM_152392.4 | NP_689605.1 | 2 - 3 | 1838 | 109 | |
GenBank mRNA | AK092505.1 | - | 2 - 3 | 225 | 109 |
AK092817.1 | - | 2 - 3 | 1242 | 109 | |
AL833813.1 | - | 2 - 3 | 1573 | 109 | |
BC063789.1 | - | 2 - 3 | 1727 | 109 | |
BC113671.1 | - | 2 - 3 | 835 | 109 | |
BC117278.1 | - | 2 - 3 | 835 | 109 |