The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015245.2 | NP_056060.2 | 14 - 15 | 2430 | 140 |
XM_005248964.3 | XP_005249021.1 | 15 - 16 | 2481 | 140 | |
XM_006715036.2 | XP_006715099.1 | 15 - 16 | 2481 | 140 | |
XM_011514431.2 | XP_011512733.1 | 15 - 16 | 2481 | 140 | |
XM_011514432.2 | XP_011512734.1 | 15 - 16 | 2481 | 140 | |
XM_011514433.1 | XP_011512735.1 | 15 - 16 | 2481 | 140 | |
XM_011514434.2 | XP_011512736.1 | 14 - 15 | 2418 | 140 | |
XM_011514435.2 | XP_011512737.1 | 14 - 15 | 2418 | 140 | |
XM_011514436.2 | XP_011512738.1 | 15 - 16 | 6050 | 140 | |
XM_011514437.2 | XP_011512739.1 | 5 - 6 | 2306 | 140 | |
XM_017010593.1 | XP_016866082.1 | 5 - 6 | 907 | 140 | |
GenBank mRNA | BC031934.1 | - | 16 - 17 | 2096 | 140 |
BC049213.1 | - | 3 - 4 | 273 | 140 | |
BC132832.1 | - | 14 - 15 | 2367 | 140 | |
D86982.1 | - | 14 - 15 | 2427 | 140 |