The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313896.1 | NP_001300825.1 | 12 - 13 | 1665 | 68 |
NM_001313982.1 | NP_001300911.1 | 12 - 13 | 1653 | 68 | |
NM_024011.3 | NP_076916.2 | 12 - 13 | 1656 | 68 | |
NM_033529.3 | NP_277071.2 | 12 - 13 | 1626 | 68 | |
NR_132739.1 | 12 - 13 | 1540 | 68 | ||
NR_132740.1 | 12 - 13 | 1579 | 68 | ||
GenBank mRNA | AF067518.1 | - | 12 - 13 | 1289 | 68 |
AF067519.1 | - | 12 - 13 | 1405 | 68 | |
AF067520.1 | - | 12 - 13 | 1402 | 68 | |
AF067521.1 | - | 12 - 13 | 1375 | 68 | |
AF067522.1 | - | 12 - 13 | 1414 | 68 | |
AF067523.1 | - | 12 - 13 | 1328 | 68 | |
AK303446.1 | - | 12 - 13 | 1414 | 68 | |
AK303451.1 | - | 12 - 13 | 1397 | 68 | |
AL043340.1 | - | 5 - 6 | 583 | 68 | |
BC014464.1 | - | 4 - 5 | 387 | 68 | |
BC094827.1 | - | 4 - 5 | 420 | 68 | |
BC110905.1 | - | 7 - 8 | 617 | 68 | |
U04819.1 | - | 5 - 6 | 729 | 68 | |
U07704.1 | - | 12 - 13 | 1437 | 68 | |
U07705.1 | - | 12 - 13 | 1431 | 68 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291345.1 | NP_001278274.1 | 12 - 13 | 1508 | 68 |
NM_001787.2 | NP_001778.2 | 12 - 13 | 1577 | 68 | |
NM_033486.2 | NP_277021.2 | 12 - 13 | 1538 | 68 | |
NM_033487.2 | NP_277022.1 | 10 - 11 | 1278 | 68 | |
NM_033489.2 | NP_277024.2 | 13 - 14 | 1585 | 68 | |
NM_033490.2 | NP_277025.1 | 11 - 12 | 1401 | 68 | |
XM_006711065.3 | XP_006711128.1 | 12 - 13 | 2289 | 68 | |
XM_011542490.2 | XP_011540792.1 | 12 - 13 | 2346 | 68 | |
XM_011542491.2 | XP_011540793.1 | 12 - 13 | 1802 | 68 | |
XM_011542492.2 | XP_011540794.1 | 12 - 13 | 1653 | 68 | |
XM_011542493.2 | XP_011540795.1 | 12 - 13 | 2316 | 68 | |
XM_011542494.2 | XP_011540796.1 | 13 - 14 | 2393 | 68 | |
XM_011542495.2 | XP_011540797.1 | 13 - 14 | 2363 | 68 | |
XM_011542496.2 | XP_011540798.1 | 7 - 8 | 833 | 68 | |
XM_017002925.1 | XP_016858414.1 | 12 - 13 | 2307 | 68 | |
XM_017002926.1 | XP_016858415.1 | 12 - 13 | 2277 | 68 | |
XM_017002927.1 | XP_016858416.1 | 12 - 13 | 2269 | 68 | |
XM_017002928.1 | XP_016858417.1 | 13 - 14 | 2366 | 68 | |
GenBank mRNA | AF067512.1 | - | 12 - 13 | 1452 | 68 |
AF067513.1 | - | 10 - 11 | 1192 | 68 | |
AF067514.1 | - | 13 - 14 | 1466 | 68 | |
AF067515.1 | - | 12 - 13 | 1491 | 68 | |
AF067516.1 | - | 13 - 14 | 1499 | 68 | |
AF067517.1 | - | 11 - 12 | 1315 | 68 | |
AF174497.1 | - | 6 - 7 | 691 | 68 | |
AK000081.1 | - | 11 - 12 | 1336 | 68 | |
AK304679.1 | - | 12 - 13 | 1416 | 68 | |
BC139777.1 | - | 6 - 7 | 621 | 68 | |
BC140714.1 | - | 12 - 13 | 1411 | 68 | |
BC171773.1 | - | 12 - 13 | 1411 | 68 | |
U04816.1 | - | 12 - 13 | 1437 | 68 | |
U04817.1 | - | 12 - 13 | 1410 | 68 | |
U04818.1 | - | 11 - 12 | 1306 | 68 | |
U04824.1 | - | 12 - 13 | 1443 | 68 |