Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5orf49
Entrez Gene ID
134121
Gene Name
chromosome 5 open reading frame 49
Gene Aliases
-
Chromosome Location
Chr.5: 7830378 - 7851490 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089584.2 NP_001083053.1 1 - 2 618 115
GenBank mRNA BC053548.1 - 1 - 2 271 115
BC062569.1 - 1 - 2 618 115
BC104204.2 - 1 - 2 279 115
BC104205.2 - 1 - 2 279 115
BC115361.1 - 1 - 2 279 115

View Full Product Details