The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024775.9 | NP_079051.9 | 1 - 2 | 98 | 71 |
GenBank mRNA | AK027112.1 | - | 1 - 2 | 68 | 71 |
BC018195.1 | - | 1 - 2 | 39 | 71 | |
BC029584.1 | - | 3 - 4 | 547 | 71 | |
BM471707.1 | - | 1 - 2 | 44 | 71 |