The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024949.5 | NP_079225.5 | 11 - 12 | 2087 | 123 |
XM_011532269.2 | XP_011530571.1 | 11 - 12 | 2720 | 123 | |
GenBank mRNA | AK126057.1 | - | 10 - 11 | 1752 | 123 |
AK127061.1 | - | 4 - 5 | 999 | 123 | |
BC107683.1 | - | 2 - 3 | 684 | 123 | |
BX647378.1 | - | 9 - 10 | 1678 | 123 |