Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148987910-148988309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC2
Entrez Gene ID
79628
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
UniGene
Hs.483784
Gene Chromosome Location
Chr.5: 148982150-149063174 on Build GRCh38
Transcripts
NM_024577.3

SNP Detail

SNP ID Chromosome Location
rs4705303 Chr.5: 148988080-148988080 on Build GRCh38
rs886715678 Chr.5: 148988245-148988245 on Build GRCh38
rs911885493 Chr.5: 148988087-148988087 on Build GRCh38
rs964495671 Chr.5: 148988005-148988005 on Build GRCh38
rs998601824 Chr.5: 148988204-148988204 on Build GRCh38
rs575957751 Chr.5: 148988141-148988141 on Build GRCh38
rs944676755 Chr.5: 148988101-148988101 on Build GRCh38
rs559757049 Chr.5: 148988238-148988238 on Build GRCh38
rs545751271 Chr.5: 148988108-148988108 on Build GRCh38
rs954361200 Chr.5: 148988030-148988030 on Build GRCh38
rs574169561 Chr.5: 148988008-148988008 on Build GRCh38
rs886060111 Chr.5: 148988204-148988204 on Build GRCh38
rs1016422956 Chr.5: 148988001-148988001 on Build GRCh38
rs141538396 Chr.5: 148988234-148988234 on Build GRCh38
rs886060109 Chr.5: 148988000-148988000 on Build GRCh38
rs983569714 Chr.5: 148988284-148988284 on Build GRCh38
rs537877706 Chr.5: 148987955-148987955 on Build GRCh38
rs564063667 Chr.5: 148988121-148988121 on Build GRCh38
rs1003247437 Chr.5: 148987973-148987973 on Build GRCh38
rs1027805686 Chr.5: 148988261-148988261 on Build GRCh38
rs187458642 Chr.5: 148988229-148988229 on Build GRCh38
rs1023179023 Chr.5: 148988236-148988236 on Build GRCh38
rs1006880290 Chr.5: 148988250-148988250 on Build GRCh38
rs1044462309 Chr.5: 148988213-148988213 on Build GRCh38
rs765288122 Chr.5: 148988281-148988281 on Build GRCh38
rs920716131 Chr.5: 148988072-148988072 on Build GRCh38
rs34395256 Chr.5: 148987935-148987935 on Build GRCh38
rs553786542 Chr.5: 148988059-148988059 on Build GRCh38
rs904293754 Chr.5: 148988181-148988181 on Build GRCh38
rs561584947 Chr.5: 148988216-148988216 on Build GRCh38
rs1041002302 Chr.5: 148988119-148988119 on Build GRCh38
rs753398688 Chr.5: 148988173-148988173 on Build GRCh38
rs138554026 Chr.5: 148987975-148987975 on Build GRCh38
rs907220974 Chr.5: 148987960-148987960 on Build GRCh38
rs993052373 Chr.5: 148987987-148987987 on Build GRCh38
rs973583665 Chr.5: 148988254-148988254 on Build GRCh38
rs1031316452 Chr.5: 148987968-148987968 on Build GRCh38
rs372760967 Chr.5: 148987988-148987988 on Build GRCh38
rs900679490 Chr.5: 148988262-148988262 on Build GRCh38
rs970278795 Chr.5: 148988239-148988239 on Build GRCh38
rs886060110 Chr.5: 148988202-148988202 on Build GRCh38
rs970200536 Chr.5: 148988024-148988024 on Build GRCh38
rs535243744 Chr.5: 148988009-148988009 on Build GRCh38
rs916409880 Chr.5: 148988120-148988120 on Build GRCh38
rs958178225 Chr.5: 148988115-148988115 on Build GRCh38
rs369963209 Chr.5: 148988149-148988149 on Build GRCh38
rs1048218251 Chr.5: 148988083-148988083 on Build GRCh38
rs916136899 Chr.5: 148987996-148987996 on Build GRCh38

View Full Product Details