The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011527214.1 | XP_011525516.1 | 3 - 4 | 427 | 136 |
XM_011527215.1 | XP_011525517.1 | 3 - 4 | 419 | 136 | |
XM_011527216.2 | XP_011525518.1 | 3 - 4 | 474 | 136 | |
GenBank mRNA | AK057404.1 | - | 3 - 4 | 419 | 136 |