The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153035.2 | NP_694580.1 | 2 - 3 | 325 | 83 |
XM_017000293.1 | XP_016855782.1 | 2 - 3 | 325 | 83 | |
GenBank mRNA | AK056674.1 | - | 2 - 3 | 296 | 83 |
BC035374.1 | - | 2 - 3 | 229 | 83 | |
DA760038.1 | - | 2 - 3 | 325 | 83 |