The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153035.2 | NP_694580.1 | 3 - 4 | 470 | 108 |
XM_017000293.1 | XP_016855782.1 | 3 - 4 | 470 | 108 | |
GenBank mRNA | AK056674.1 | - | 3 - 4 | 441 | 108 |
BC035374.1 | - | 3 - 4 | 374 | 108 |