Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNHD1
Entrez Gene ID
144132
Gene Name
dynein heavy chain domain 1
Gene Aliases
C11orf47, CCDC35, DHCD1, DNHD1L
Chromosome Location
Chr.11: 6497296 - 6572025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144666.2 NP_653267.2 6 - 7 1800 69
NM_173589.3 NP_775860.3 5 - 6 1903 69
GenBank mRNA AK093028.1 - 2 - 3 984 69
BC111765.1 - 4 - 5 1653 69
BX648742.1 - 5 - 6 1898 69

View Full Product Details