Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC9A9
Entrez Gene ID
285195
Gene Name
solute carrier family 9 member A9
Gene Aliases
AUTS16, NHE9
Chromosome Location
Chr.3: 143265222 - 143848531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173653.3 NP_775924.1 10 - 11 1418 121
XM_011512703.2 XP_011511005.1 7 - 8 728 121
XM_017006202.1 XP_016861691.1 10 - 11 1391 121
XM_017006203.1 XP_016861692.1 9 - 10 1176 121
GenBank mRNA AB089794.1 - 10 - 11 1209 121
AK092932.1 - 4 - 5 469 121
AK315107.1 - 10 - 11 1345 121
AL832304.1 - 10 - 11 1283 121
AY254100.1 - 10 - 11 1333 121
BC035779.1 - 10 - 11 1334 121

View Full Product Details