The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173653.3 | NP_775924.1 | 15 - 16 | 1918 | 73 |
XM_011512703.2 | XP_011511005.1 | 12 - 13 | 1228 | 73 | |
XM_017006203.1 | XP_016861692.1 | 14 - 15 | 1676 | 73 | |
GenBank mRNA | AB075486.1 | - | 5 - 6 | 419 | 73 |
AB089794.1 | - | 15 - 16 | 1709 | 73 | |
AK092932.1 | - | 9 - 10 | 969 | 73 | |
AK315107.1 | - | 15 - 16 | 1845 | 73 | |
AL832304.1 | - | 15 - 16 | 1783 | 73 | |
AY254100.1 | - | 15 - 16 | 1833 | 73 | |
BC035779.1 | - | 15 - 16 | 1834 | 73 |