The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174976.2 | NP_777636.2 | 1 - 2 | 190 | 77 |
GenBank mRNA | AK095612.1 | - | 1 - 2 | 190 | 77 |
AK294840.1 | - | 1 - 2 | 189 | 77 | |
DA496626.1 | - | 1 - 2 | 190 | 77 |