The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175857.3 | NP_787053.1 | 1 - 1 | 178 | 60 |
GenBank mRNA | AB096961.1 | - | 1 - 1 | 176 | 60 |
AJ457064.1 | - | 1 - 1 | 178 | 60 | |
BC130397.1 | - | 1 - 1 | 169 | 60 | |
BC133033.1 | - | 1 - 1 | 169 | 60 |