Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDFY1
Entrez Gene ID
57590
Gene Name
WD repeat and FYVE domain containing 1
Gene Aliases
FENS-1, WDF1, ZFYVE17
Chromosome Location
Chr.2: 223875343 - 223945387 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020830.4 NP_065881.1 6 - 7 702 81
GenBank mRNA AB037856.1 - 6 - 7 615 81
AF411977.1 - 6 - 7 599 81
AJ310568.1 - 6 - 7 689 81
AK022888.1 - 6 - 7 628 81
AK023415.1 - 5 - 6 496 81
BC040525.1 - 6 - 7 614 81
BC065934.1 - 6 - 7 646 81

View Full Product Details