The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031369.2 | NP_112737.1 | 6 - 7 | 1267 | 137 |
NM_031370.2 | NP_112738.1 | 7 - 8 | 1324 | 137 | |
GenBank mRNA | AK057836.1 | - | 7 - 8 | 1285 | 137 |
AK292707.1 | - | 6 - 7 | 1262 | 137 | |
AK300149.1 | - | 7 - 8 | 1156 | 137 | |
AK303552.1 | - | 5 - 6 | 1093 | 137 | |
BC002401.1 | - | 7 - 8 | 1070 | 137 | |
D55671.1 | - | 6 - 7 | 743 | 137 | |
D55674.1 | - | 7 - 8 | 1270 | 137 |