Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL5A1
Entrez Gene ID
1289
Gene Name
collagen type V alpha 1
Gene Aliases
EDSC
Chromosome Location
Chr.9: 134641790 - 134844843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000093.4 NP_000084.3 8 - 9 1720 68
NM_001278074.1 NP_001265003.1 8 - 9 1720 68
XM_017014266.1 XP_016869755.1 8 - 9 1735 68
GenBank mRNA AB209864.1 - 7 - 8 1199 68
AB371583.1 - 8 - 9 1720 68
CN399221.1 - 2 - 3 239 68
D90279.1 - 8 - 9 1463 68

View Full Product Details