The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014809.2 | NP_001014809.1 | 13 - 14 | 2089 | 58 |
NM_001288661.1 | NP_001275590.1 | 13 - 14 | 2196 | 58 | |
NM_001288662.1 | NP_001275591.1 | 13 - 14 | 2019 | 58 | |
NM_001313.4 | NP_001304.1 | 13 - 14 | 1983 | 58 | |
GenBank mRNA | AK091696.1 | - | 1 - 2 | 1367 | 58 |
AK094725.1 | - | 13 - 14 | 1748 | 58 | |
AK122754.1 | - | 13 - 14 | 2019 | 58 | |
AK127754.1 | - | 14 - 15 | 1735 | 58 | |
AK309379.1 | - | - | 438 | 58 | |
BC000252.1 | - | 13 - 14 | 1848 | 58 | |
BC007613.1 | - | 13 - 14 | 1848 | 58 | |
BC007898.2 | - | 9 - 10 | 1131 | 58 | |
BT006806.1 | - | 13 - 14 | 1633 | 58 | |
D78012.1 | - | 13 - 14 | 1783 | 58 | |
DQ206871.1 | - | 13 - 14 | 1975 | 58 | |
KJ534816.1 | - | 13 - 14 | 1975 | 58 | |
KJ534817.1 | - | 14 - 15 | 1882 | 58 | |
U17278.1 | - | 11 - 12 | 1460 | 58 |