Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18785962-18786189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMP
Entrez Gene ID
1311
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
UniGene
Hs.1584
Gene Chromosome Location
Chr.19: 18782773-18791305 on Build GRCh38
Transcripts
NM_000095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5601300 c.1432C>T p.P478S Chr.19: 18786022-18786022 on Build GRCh38
310278 c.1424A>T p.D475V Chr.19: 18786030-18786030 on Build GRCh38
993159 c.1406A>G p.D469G Chr.19: 18786048-18786048 on Build GRCh38
417690 c.1475A>T p.Q492L Chr.19: 18785979-18785979 on Build GRCh38
460016 c.1462C>T p.P488S Chr.19: 18785992-18785992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761508334 Chr.19: 18786047-18786047 on Build GRCh38
rs747106682 Chr.19: 18786123-18786123 on Build GRCh38
rs137852651 Chr.19: 18786051-18786051 on Build GRCh38
rs312262897 Chr.19: 18786035-18786037 on Build GRCh38
rs28936668 Chr.19: 18786096-18786096 on Build GRCh38
rs774266370 Chr.19: 18786141-18786141 on Build GRCh38
rs762683869 Chr.19: 18786013-18786013 on Build GRCh38
rs749899512 Chr.19: 18786161-18786161 on Build GRCh38
rs191645921 Chr.19: 18786001-18786001 on Build GRCh38
rs759742248 Chr.19: 18786152-18786154 on Build GRCh38
rs368273443 Chr.19: 18786137-18786137 on Build GRCh38
rs28936669 Chr.19: 18786036-18786036 on Build GRCh38
rs751147399 Chr.19: 18786044-18786044 on Build GRCh38
rs755277222 Chr.19: 18786079-18786079 on Build GRCh38
rs530668887 Chr.19: 18786102-18786102 on Build GRCh38
rs375031149 Chr.19: 18786050-18786050 on Build GRCh38
rs775306610 Chr.19: 18786012-18786012 on Build GRCh38
rs769866414 Chr.19: 18786008-18786008 on Build GRCh38
rs761399284 Chr.19: 18786147-18786147 on Build GRCh38
rs543419638 Chr.19: 18786107-18786107 on Build GRCh38
rs766950514 Chr.19: 18786048-18786048 on Build GRCh38
rs28933699 Chr.19: 18786043-18786043 on Build GRCh38
rs773490189 Chr.19: 18785991-18785991 on Build GRCh38
rs768883872 Chr.19: 18785984-18785984 on Build GRCh38
rs756568697 Chr.19: 18786055-18786055 on Build GRCh38
rs541996591 Chr.19: 18785983-18785983 on Build GRCh38
rs777939991 Chr.19: 18786120-18786120 on Build GRCh38
rs772268073 Chr.19: 18785987-18785987 on Build GRCh38
rs754409266 Chr.19: 18786078-18786078 on Build GRCh38
rs950287971 Chr.19: 18786160-18786160 on Build GRCh38
rs895998694 Chr.19: 18786121-18786121 on Build GRCh38
rs375476152 Chr.19: 18786070-18786070 on Build GRCh38
rs867983497 Chr.19: 18785986-18785986 on Build GRCh38
rs763986060 Chr.19: 18786021-18786021 on Build GRCh38
rs137852650 Chr.19: 18786040-18786040 on Build GRCh38
rs775255655 Chr.19: 18786129-18786129 on Build GRCh38
rs868493730 Chr.19: 18785997-18785997 on Build GRCh38
rs765564022 Chr.19: 18786157-18786160 on Build GRCh38
rs1045068010 Chr.19: 18786009-18786009 on Build GRCh38
rs762992681 Chr.19: 18786130-18786130 on Build GRCh38
rs779394384 Chr.19: 18786099-18786099 on Build GRCh38
rs370788977 Chr.19: 18786150-18786150 on Build GRCh38
rs1056333727 Chr.19: 18786101-18786101 on Build GRCh38
rs780461514 Chr.19: 18786066-18786066 on Build GRCh38
rs865949126 Chr.19: 18785998-18785998 on Build GRCh38
rs748431274 Chr.19: 18785985-18785985 on Build GRCh38
rs193922900 Chr.19: 18786035-18786037 on Build GRCh38
rs312262898 Chr.19: 18786034-18786035 on Build GRCh38

View Full Product Details