Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS3
Entrez Gene ID
9508
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 3
Gene Aliases
ADAMTS-4
Chromosome Location
Chr.4: 72280969 - 72568799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014243.2 NP_055058.2 21 - 22 3084 85
XM_011532421.2 XP_011530723.1 21 - 22 3116 85
XM_011532422.2 XP_011530724.1 21 - 22 3615 85
GenBank mRNA AB002364.1 - 21 - 22 3036 85
BC016451.1 - 4 - 5 524 85
BC130287.1 - 21 - 22 3081 85
BC132735.1 - 21 - 22 3081 85

View Full Product Details