Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16989518-16989765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3849324 c.2592C>T p.C864C Chr.1: 16989708-16989708 on Build GRCh38
3849325 c.2460C>T p.C820C Chr.1: 16989708-16989708 on Build GRCh38
7324241 c.2593G>A p.E865K Chr.1: 16989707-16989707 on Build GRCh38
7356921 c.2437G>T p.E813* Chr.1: 16989731-16989731 on Build GRCh38
7356920 c.2569G>T p.E857* Chr.1: 16989731-16989731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563317561 Chr.1: 16989654-16989654 on Build GRCh38
rs751168600 Chr.1: 16989680-16989680 on Build GRCh38
rs140118225 Chr.1: 16989573-16989573 on Build GRCh38
rs752494943 Chr.1: 16989701-16989701 on Build GRCh38
rs374091614 Chr.1: 16989634-16989634 on Build GRCh38
rs910333657 Chr.1: 16989575-16989575 on Build GRCh38
rs753813388 Chr.1: 16989719-16989719 on Build GRCh38
rs886045579 Chr.1: 16989737-16989737 on Build GRCh38
rs947121579 Chr.1: 16989577-16989577 on Build GRCh38
rs376194683 Chr.1: 16989655-16989655 on Build GRCh38
rs747242869 Chr.1: 16989746-16989746 on Build GRCh38
rs765905631 Chr.1: 16989675-16989675 on Build GRCh38
rs905316433 Chr.1: 16989637-16989637 on Build GRCh38
rs587776890 Chr.1: 16989747-16989748 on Build GRCh38
rs1042911063 Chr.1: 16989586-16989586 on Build GRCh38
rs530094467 Chr.1: 16989603-16989603 on Build GRCh38
rs745954026 Chr.1: 16989742-16989742 on Build GRCh38
rs761130666 Chr.1: 16989648-16989648 on Build GRCh38
rs954861510 Chr.1: 16989656-16989656 on Build GRCh38
rs148534162 Chr.1: 16989707-16989707 on Build GRCh38
rs749694399 Chr.1: 16989622-16989622 on Build GRCh38
rs778906228 Chr.1: 16989740-16989740 on Build GRCh38
rs777472886 Chr.1: 16989708-16989708 on Build GRCh38
rs1045067591 Chr.1: 16989636-16989636 on Build GRCh38
rs924440516 Chr.1: 16989592-16989592 on Build GRCh38
rs587777053 Chr.1: 16989739-16989739 on Build GRCh38
rs905170502 Chr.1: 16989610-16989610 on Build GRCh38
rs772243999 Chr.1: 16989743-16989743 on Build GRCh38
rs373475292 Chr.1: 16989646-16989646 on Build GRCh38
rs1023513011 Chr.1: 16989576-16989576 on Build GRCh38
rs55889979 Chr.1: 16989744-16989744 on Build GRCh38
rs772297257 Chr.1: 16989643-16989643 on Build GRCh38
rs759204814 Chr.1: 16989681-16989681 on Build GRCh38
rs370828598 Chr.1: 16989660-16989660 on Build GRCh38
rs369871753 Chr.1: 16989633-16989633 on Build GRCh38
rs767280251 Chr.1: 16989700-16989700 on Build GRCh38
rs376174719 Chr.1: 16989638-16989638 on Build GRCh38
rs774420241 Chr.1: 16989736-16989736 on Build GRCh38

View Full Product Details