The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204830.1 | NP_001191759.1 | 3 - 3 | 659 | 153 |
NM_015929.3 | NP_057013.1 | 3 - 3 | 461 | 153 | |
NM_145197.2 | NP_660198.1 | 4 - 4 | 589 | 153 | |
NM_145198.2 | NP_660199.1 | 4 - 4 | 577 | 153 | |
NM_145199.2 | NP_660200.1 | 2 - 2 | 335 | 153 | |
NR_037935.1 | 4 - 4 | 722 | 153 | ||
NR_037936.1 | 3 - 3 | 398 | 153 | ||
GenBank mRNA | AB017566.1 | - | 3 - 3 | 461 | 153 |
BC007001.1 | - | 3 - 3 | 381 | 153 | |
BC009772.1 | - | 3 - 3 | 402 | 153 | |
BC070145.1 | - | 3 - 3 | 439 | 153 | |
BQ422048.1 | - | 3 - 3 | 370 | 153 |