Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37876751-37876978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6490277 c.935G>A p.S312N Chr.7: 37876948-37876948 on Build GRCh38
3229117 c.893A>C p.K298T Chr.7: 37876906-37876906 on Build GRCh38
7379894 c.851C>A p.A284D Chr.7: 37876864-37876864 on Build GRCh38
3638522 c.912C>T p.F304F Chr.7: 37876925-37876925 on Build GRCh38
601158 c.874G>A p.D292N Chr.7: 37876887-37876887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755364633 Chr.7: 37876907-37876907 on Build GRCh38
rs202155606 Chr.7: 37876869-37876869 on Build GRCh38
rs952358772 Chr.7: 37876837-37876837 on Build GRCh38
rs756633731 Chr.7: 37876880-37876880 on Build GRCh38
rs138867361 Chr.7: 37876842-37876842 on Build GRCh38
rs750935741 Chr.7: 37876879-37876879 on Build GRCh38
rs775303606 Chr.7: 37876854-37876854 on Build GRCh38
rs763550235 Chr.7: 37876872-37876872 on Build GRCh38
rs770715891 Chr.7: 37876938-37876938 on Build GRCh38
rs758445291 Chr.7: 37876929-37876929 on Build GRCh38
rs763443385 Chr.7: 37876853-37876855 on Build GRCh38
rs777785559 Chr.7: 37876930-37876930 on Build GRCh38
rs776349211 Chr.7: 37876805-37876805 on Build GRCh38
rs766162606 Chr.7: 37876910-37876910 on Build GRCh38
rs146004225 Chr.7: 37876928-37876928 on Build GRCh38
rs779061373 Chr.7: 37876926-37876926 on Build GRCh38
rs1026715463 Chr.7: 37876827-37876827 on Build GRCh38
rs570286756 Chr.7: 37876919-37876919 on Build GRCh38
rs764411137 Chr.7: 37876934-37876935 on Build GRCh38
rs754132765 Chr.7: 37876897-37876897 on Build GRCh38
rs776637215 Chr.7: 37876942-37876942 on Build GRCh38
rs62001868 Chr.7: 37876853-37876853 on Build GRCh38
rs774993830 Chr.7: 37876808-37876808 on Build GRCh38
rs201707476 Chr.7: 37876794-37876794 on Build GRCh38
rs770757133 Chr.7: 37876791-37876791 on Build GRCh38
rs746954043 Chr.7: 37876934-37876934 on Build GRCh38
rs766918975 Chr.7: 37876895-37876895 on Build GRCh38

View Full Product Details