Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CATIP
Entrez Gene ID
375307
Gene Name
ciliogenesis associated TTC17 interacting protein
Gene Aliases
C2orf62
Chromosome Location
Chr.2: 218356757 - 218368393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320865.1 NP_001307794.1 8 - 9 896 107
NM_198559.1 NP_940961.1 8 - 9 863 107
XM_005246539.4 XP_005246596.1 7 - 8 860 107
XM_005246541.4 XP_005246598.1 6 - 7 659 107
XM_011511148.2 XP_011509450.1 7 - 8 827 107
XM_011511149.2 XP_011509451.1 6 - 7 658 107
XM_011511150.2 XP_011509452.1 6 - 7 624 107
XM_017004053.1 XP_016859542.1 6 - 7 1059 107
GenBank mRNA BC052750.1 - 8 - 9 863 107

View Full Product Details