The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005261.3 | NP_005252.1 | 4 - 5 | 857 | 69 |
NM_181702.2 | NP_859053.1 | 4 - 5 | 742 | 69 | |
GenBank mRNA | AK314017.1 | - | 4 - 5 | 742 | 69 |
BC022010.2 | - | 4 - 5 | 742 | 69 | |
U10550.1 | - | 4 - 5 | 831 | 69 | |
U13052.1 | - | 3 - 4 | 618 | 69 |