The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004537.5 | NP_004528.1 | 15 - 15 | 3125 | 139 |
NM_139207.3 | NP_631946.1 | 16 - 16 | 2451 | 139 | |
XM_006719425.3 | XP_006719488.1 | 14 - 14 | 1914 | 139 | |
XM_011538390.2 | XP_011536692.2 | 16 - 16 | 2082 | 139 | |
XM_011538393.2 | XP_011536695.1 | 16 - 16 | 2363 | 139 | |
XM_017019338.1 | XP_016874827.1 | 16 - 16 | 2335 | 139 | |
XM_017019339.1 | XP_016874828.1 | 14 - 14 | 2174 | 139 | |
XM_017019340.1 | XP_016874829.1 | 14 - 14 | 1886 | 139 | |
GenBank mRNA | AF300335.2 | - | 1 - 1 | 277 | 139 |
AL162068.1 | - | 15 - 15 | 2765 | 139 |