The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021992.2 | NP_068832.1 | 3 - 3 | 398 | 116 |
GenBank mRNA | AF086495.1 | - | 3 - 3 | 341 | 116 |
BC000183.1 | - | 3 - 3 | 346 | 116 | |
BG829126.1 | - | 3 - 3 | 348 | 116 | |
BU599402.1 | - | 3 - 3 | 411 | 116 | |
D82345.1 | - | 3 - 3 | 372 | 116 |