Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF77
Entrez Gene ID
58492
Gene Name
zinc finger protein 77
Gene Aliases
pT1
Chromosome Location
Chr.19: 2933218 - 2944971 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021217.2 NP_067040.1 2 - 3 265 73
XM_017027082.1 XP_016882571.1 2 - 3 290 108
GenBank mRNA AK290979.1 - 2 - 3 201 73
BC043354.1 - 2 - 3 226 73
BC062561.1 - 2 - 3 227 73

View Full Product Details