Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS1A
Entrez Gene ID
23294
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
Gene Aliases
ANKS1
Chromosome Location
Chr.6: 34889261 - 35097994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015245.2 NP_056060.2 23 - 24 3546 68
XM_005248964.3 XP_005249021.1 24 - 25 3597 68
XM_006715036.2 XP_006715099.1 24 - 25 3600 68
XM_017010593.1 XP_016866082.1 14 - 15 2023 68
GenBank mRNA BC049213.1 - 12 - 13 1389 68
BC132832.1 - 23 - 24 3483 68
BX094969.1 - 6 - 7 675 68
D86982.1 - 23 - 24 3543 68

View Full Product Details