The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001093770.2 | NP_001087239.2 | 6 - 6 | 1538 | 164 |
NM_001164644.1 | NP_001158116.1 | 6 - 6 | 1508 | 164 | |
NM_001164645.1 | NP_001158117.1 | 6 - 6 | 1394 | 164 | |
NM_001164646.1 | NP_001158118.1 | 6 - 6 | 1402 | 164 | |
NM_001164647.1 | NP_001158119.1 | 5 - 5 | 1478 | 164 | |
NM_005411.4 | NP_005402.3 | 6 - 6 | 1549 | 164 | |
XM_005270062.4 | XP_005270119.1 | 5 - 5 | 1538 | 164 | |
XM_006717953.2 | XP_006718016.1 | 6 - 6 | 1531 | 164 | |
GenBank mRNA | AK298029.1 | - | 5 - 5 | 1438 | 164 |
BC026229.1 | - | 2 - 2 | 1021 | 164 | |
HQ021433.2 | - | 5 - 5 | 1478 | 164 | |
HQ021434.1 | - | 5 - 5 | 1489 | 164 | |
HQ021435.1 | - | 5 - 5 | 1478 | 164 | |
HQ021436.1 | - | 5 - 5 | 1477 | 164 | |
HQ021437.1 | - | 6 - 6 | 1560 | 164 | |
HQ021438.1 | - | 6 - 6 | 1549 | 164 | |
HQ021439.1 | - | 6 - 6 | 1548 | 164 | |
HQ021440.1 | - | 6 - 6 | 1549 | 164 | |
HQ021441.1 | - | 6 - 6 | 1538 | 164 | |
HQ021442.1 | - | 6 - 6 | 1537 | 164 | |
JX502764.1 | - | 6 - 6 | 1549 | 164 | |
JX502765.1 | - | 6 - 6 | 1538 | 164 |