The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053055.4 | NP_444283.2 | 6 - 6 | 1807 | 163 |
XM_011509142.2 | XP_011507444.1 | 7 - 7 | 1465 | 163 |