Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
Chromosome Location
Chr.11: 35431827 - 35530300 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001991.2 NP_001001991.1 2 - 3 695 76
NM_001282675.1 NP_001269604.1 4 - 5 685 76
NM_001282676.1 NP_001269605.1 2 - 3 695 76
NM_015430.3 NP_056245.2 2 - 3 695 76
GenBank mRNA AF370388.1 - 1 - 2 183 76
AK027841.1 - 2 - 3 295 76
AK297092.1 - 4 - 5 686 76
AK300185.1 - 2 - 3 280 76
AK316188.1 - 2 - 3 267 76
AL832391.1 - 2 - 3 261 76
AY358346.1 - 2 - 3 274 76
BC089434.1 - 2 - 3 287 76
BX640676.1 - 2 - 3 695 76

View Full Product Details