The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284262.1 | NP_001271191.1 | 3 - 4 | 678 | 82 |
NM_198285.2 | NP_938026.2 | 3 - 4 | 1177 | 82 | |
XM_005249990.4 | XP_005250047.1 | 3 - 4 | 1174 | 82 | |
XM_006715966.2 | XP_006716029.1 | 3 - 4 | 1174 | 82 | |
XM_011516146.2 | XP_011514448.1 | 3 - 4 | 1174 | 82 | |
GenBank mRNA | BC107125.2 | - | 2 - 3 | 574 | 82 |
BX390499.1 | - | 1 - 2 | 284 | 82 |