Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRT4
Entrez Gene ID
401399
Gene Name
proline rich transmembrane protein 4
Gene Aliases
-
Chromosome Location
Chr.7: 128350325 - 128362067 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114726.2 NP_001108198.2 1 - 2 128 89
XM_005250343.3 XP_005250400.1 1 - 2 154 89

View Full Product Details