Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCLC
Entrez Gene ID
2729
Gene Name
glutamate-cysteine ligase catalytic subunit
Gene Aliases
GCL, GCS, GLCL, GLCLC
Chromosome Location
Chr.6: 53497341 - 53545129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197115.1 NP_001184044.1 13 - 14 1948 74
NM_001498.3 NP_001489.1 14 - 15 2062 74
XM_017010749.1 XP_016866238.1 14 - 15 1687 74
GenBank mRNA AB262176.1 - 14 - 15 1965 74
AB516284.1 - 13 - 14 1464 74
AK094940.1 - 2 - 3 74
AK304288.1 - 14 - 15 1590 74
AK313480.1 - 14 - 15 1972 74
BC039894.1 - 14 - 15 1660 74
M90656.1 - 14 - 15 1670 74

View Full Product Details