Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GMEB1
Entrez Gene ID
10691
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
P96PIF, PIF96
Chromosome Location
Chr.1: 28668728 - 28715603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319674.1 NP_001306603.1 9 - 10 1134 76
NM_006582.3 NP_006573.2 9 - 10 1164 76
NM_024482.2 NP_077808.1 9 - 10 1131 76
XM_011540518.2 XP_011538820.1 9 - 10 1267 76
XM_011540519.2 XP_011538821.1 9 - 10 1107 76
XM_011540521.2 XP_011538823.1 9 - 10 1236 76
XM_011540522.2 XP_011538824.1 9 - 10 1083 76
XM_017000087.1 XP_016855576.1 9 - 10 1282 76
GenBank mRNA AF099013.1 - 8 - 9 1028 76
AF173868.1 - 9 - 10 1059 76
AF203694.1 - 9 - 10 1058 76
AK000892.1 - 9 - 10 1130 76
AK315436.1 - 9 - 10 1068 76
BC001473.2 - 9 - 10 1081 76

View Full Product Details