The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167599.2 | NP_001161071.1 | 2 - 3 | 741 | 131 |
NM_001167600.2 | NP_001161072.1 | 2 - 3 | 418 | 131 | |
NM_001167601.2 | NP_001161073.1 | 2 - 3 | 421 | 131 | |
NM_001167602.2 | NP_001161074.1 | 3 - 4 | 919 | 131 | |
NM_080741.3 | NP_542779.2 | 2 - 3 | 738 | 131 | |
GenBank mRNA | AJ277883.1 | - | 1 - 2 | 205 | 131 |
AK025617.1 | - | 2 - 3 | 290 | 131 | |
AK091038.1 | - | 3 - 4 | 916 | 131 | |
AK289421.1 | - | 2 - 3 | 295 | 131 | |
AK290076.1 | - | 2 - 3 | 278 | 131 | |
AK298403.1 | - | 3 - 4 | 250 | 131 | |
AK300969.1 | - | 2 - 3 | 291 | 131 | |
AY203952.1 | - | 2 - 3 | 274 | 131 | |
BC012899.1 | - | 2 - 3 | 692 | 131 | |
BC105974.1 | - | 2 - 3 | 254 | 131 | |
BC117214.1 | - | 2 - 3 | 382 | 131 |