Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP2
Entrez Gene ID
9099
Gene Name
ubiquitin specific peptidase 2
Gene Aliases
UBP41, USP9
Chromosome Location
Chr.11: 119355215 - 119381726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243759.1 NP_001230688.1 7 - 8 824 73
NM_004205.4 NP_004196.4 8 - 9 1639 73
NM_171997.2 NP_741994.1 7 - 8 904 73
XM_005271721.4 XP_005271778.1 8 - 9 1512 73
XM_005271722.2 XP_005271779.1 8 - 9 1518 73
XM_017018539.1 XP_016874028.1 7 - 8 600 73
GenBank mRNA AF079564.1 - 7 - 8 642 73
AF440755.1 - 7 - 8 904 73
AK057225.1 - 8 - 9 1568 73
AK292255.1 - 7 - 8 827 73
AK301805.1 - 7 - 8 709 73
BC002854.1 - 8 - 9 1520 73
BC002955.1 - 8 - 9 1520 73
BC041366.2 - 7 - 8 809 73

View Full Product Details