Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SESN2
Entrez Gene ID
83667
Gene Name
sestrin 2
Gene Aliases
HI95, SES2, SEST2
Chromosome Location
Chr.1: 28259452 - 28282491 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031459.4 NP_113647.1 4 - 5 932 55
GenBank mRNA AK027896.1 - 2 - 3 245 55
AK315710.1 - 4 - 5 645 55
AL136551.1 - 4 - 5 712 55
AY123223.1 - 4 - 5 858 55
BC013304.2 - 4 - 5 860 55
BC033719.1 - 4 - 5 866 55

View Full Product Details