The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031459.4 | NP_113647.1 | 5 - 6 | 1147 | 50 |
GenBank mRNA | AK027896.1 | - | 3 - 4 | 460 | 50 |
AK315710.1 | - | 5 - 6 | 860 | 50 | |
AL136551.1 | - | 5 - 6 | 927 | 50 | |
AY123223.1 | - | 5 - 6 | 1073 | 50 | |
BC013304.2 | - | 5 - 6 | 1075 | 50 | |
BC033719.1 | - | 5 - 6 | 1081 | 50 |