The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031459.4 | NP_113647.1 | 6 - 7 | 1300 | 94 |
GenBank mRNA | AK027896.1 | - | 4 - 5 | 613 | 94 |
AK315710.1 | - | 6 - 7 | 1013 | 94 | |
AL136551.1 | - | 6 - 7 | 1080 | 94 | |
AY123223.1 | - | 6 - 7 | 1226 | 94 | |
BC013304.2 | - | 6 - 7 | 1228 | 94 | |
BC033719.1 | - | 6 - 7 | 1234 | 94 |